Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • c-Met Antibodies

          Invitrogen

          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Polyclonal Antibody

          15 Published Figures
          39 References
          View all (98) c-Met antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (15)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 17

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody (44-888G) in WB

          Peptide Competition: Extracts prepared from 293T cells left unstimulated (1) or stimulated with HGF (2-5) were resolved by SDS-PAGE on a 10% polyacrylamide gel and transferred to PVDF. Membranes were blocked with a 5% BSA-TBST buffer overnight at 4°C {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Western Blot (WB)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Immunohistochemistry (IHC)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Immunohistochemistry (IHC)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Immunohistochemistry (IHC)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Immunohistochemistry (IHC)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Immunohistochemistry (IHC)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Immunohistochemistry (IHC)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Immunohistochemistry (IHC)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Immunohistochemistry (IHC)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Western Blot (WB)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Western Blot (WB)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Western Blot (WB)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Western Blot (WB)
          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Polyclonal Antibody

          Product Details

          44-888G

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 20 publications 20 publications

          Immunohistochemistry (IHC)

          -
          View 8 publications 8 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 6 publications 6 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Dog, Hamster, Human, Mouse, Rat, Virus

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human c-Met that contains tyrosines 1230, 1234 and 1235. The sequence is conserved in mouse and rat.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Antigen affinity chromatography

          Storage buffer

          Dulbecco's PBS, pH 7.3, with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533787

          Product Specific Information

          44-888G has been used successfully in peptide-ELISA. Dilution is to be determined by the consumer.

          Target Information

          MET (cMET) is a receptor-like tyrosine kinase whose dysregulation has been linked to many types of human malignancies. After activation of the ligand, MET interacts with the PI3-kinase subunit PIK3R1, PLCG1, SRC, GRB2, and STAT3. There interactions lead to the activation of signaling cascades including RAS-ERK, PI3, kinase-AKT, and PLCgamma-PKC. MET plays a role in embryonic development including gastrulation, development of muscles and neurons, angiogenesis, and kidney formation. It also plays a role in adults including wound healing, organ regeneration, and tissue remodeling. MET has been linked to cancers including gastric, renal, and breast; therefore, making it a target for cancer therapeutics and diagnostic testing.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cMet; EC 2.7.10.1; HGF receptor; HGF-SF receptor; HGF/SF receptor; met oncogene; MET proto-oncogene protein; met proto-oncogene tyrosine kinase; oncogene MET; protein-tyrosine kinase (EC 2.7.1.112); proto-oncogene c-Met; sc MET; scatter factor receptor; SF receptor; soluble c met; tyrosine kinase; hepatocyte growth factor receptor; HGFR; tyrosine-protein kinase Met; unnamed protein product

          View more View less

          Gene Aliases: AUTS9; c-Met; DA11; DFNB97; HGF; HGFR; Par4; RCCP2

          View more View less

          UniProt ID: (Human) P08581

          View more View less

          Entrez Gene ID: (Human) 4233, (Mouse) 17295

          View more View less

          Function(s)
          protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity hepatocyte growth factor-activated receptor activity protein binding ATP binding semaphorin receptor activity protein phosphatase binding identical protein binding molecular function activator activity beta-catenin binding phospholipase binding phosphatidylinositol 3-kinase binding macromolecular complex binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter neuron migration endothelial cell morphogenesis liver development placenta development transmembrane receptor protein tyrosine kinase signaling pathway chemical synaptic transmission brain development muscle organ development skeletal muscle tissue development regulation of gene expression positive regulation of gene expression negative regulation of gene expression positive regulation of peptidyl-threonine phosphorylation negative regulation of peptidyl-threonine phosphorylation positive regulation of glucose transport positive regulation of neuron projection development muscle cell migration myotube differentiation regulation of anatomical structure morphogenesis neuron differentiation sperm motility adult behavior pancreas development positive regulation of microtubule polymerization regulation of interleukin-6 production positive regulation of peptidyl-serine phosphorylation negative regulation of Rho protein signal transduction glucose homeostasis positive regulation of MAPK cascade positive regulation of DNA replication positive regulation of mitotic nuclear division positive regulation of transcription from RNA polymerase II promoter protein autophosphorylation hepatocyte growth factor receptor signaling pathway cell development branching morphogenesis of an epithelial tube positive regulation of dendrite morphogenesis modulation of synaptic transmission positive chemotaxis myoblast proliferation negative regulation of stress fiber assembly cardiac muscle cell development cardiac muscle contraction excitatory postsynaptic potential regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling establishment of skin barrier negative regulation of thrombin receptor signaling pathway semaphorin-plexin signaling pathway negative regulation of transforming growth factor beta production reactive oxygen species metabolic process regulation of cellular response to oxidative stress positive regulation of p38MAPK cascade negative regulation of hydrogen peroxide-mediated programmed cell death positive regulation of endothelial cell chemotaxis cell surface receptor signaling pathway negative regulation of autophagy
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch