DNA Binding Beads for MagMAX™-96 DNA Multi-Sample Kit - FAQs
DNA Binding Beads for MagMAX™-96 DNA Multi-Sample Kit - FAQs
View additional product information for DNA Binding Beads for MagMAX™-96 DNA Multi-Sample Kit - FAQs (4489112)
1 product FAQs found
Can I purchase the RNA Binding beads from the MagMAX MirVana Total RNA Isolation Kit (Cat. No. A27828) separately?
Yes, you can. The DNA Binding Beads for MagMAX-96 DNA Multi-Sample Kit (Cat. No. 4489112) are the same as that used in A27828.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.