Pierce™ Conjugate Purification Kit
Thermo Scientific™

Pierce™ Conjugate Purification Kit

The Thermo Scientific Pierce Conjugate Purification Kit is a gentle affinity chromatography system for purifying horseradish peroxidase (HRP) or alkalineRead more
Have Questions?
Catalog NumberQuantity
449201 Kit
Catalog number 44920
Price (USD)
828.00
Each
Quantity:
1 Kit
Request bulk or custom format
Price (USD)
828.00
Each
The Thermo Scientific Pierce Conjugate Purification Kit is a gentle affinity chromatography system for purifying horseradish peroxidase (HRP) or alkaline phosphatase (AP) conjugated antibodies from unreacted enzyme after labeling.

Features of the Conjugate Purification Kit:

• Specific for IgG—procedure is designed for removal of HRP or AP enzymes from IgG antibodies; not for use with IgM, IgY, Fab fragments or streptavidin conjugates
• Complete kit—includes affinity column, activator and elution buffers, desalting column and storage buffers for HRP and AP conjugates
• High capacity—the single-use column-kit is designed for purification of 0.5 to 50 mg of antibody conjugate dissolved in Tris-buffered saline
• Improves assays—removing unconjugated enzyme from antibody conjugates often reduces background and improves overal performance of immunoassays

This conjugate purification kit is designed to eliminate excess, unconjugated HRP or Alk-Phos enzyme from labeled antibodies (IgG) after performing antibody-enzyme conjugation reactions. The column of metal-chelate agarose resin binds to IgG-enzyme conjugates but allows non-conjugated enzyme (HRP or AP) to to pass through. A gentle, non-denaturing elution buffer recovers the fully active IgG-HRP or IgG-AP conjugate. The included desalting columns exchange the purified conjugate into any buffer of choice, ready for use in various immunodetection methods.

In this kit procedure, the nickel-chelated agarose binds IgG through a histidine-rich cluster on the Fc region at the junctures of the Cγ2 and Cγ3 domains, which is highly conserved across all mammalian IgGs. Purified IgG from sheep, mouse, goat, rat and rabbit will bind to nickel-chelated resin. Avidin or streptavidin enzyme conjugates cannot be purified by this method because they do not bind to the nickel-chelated agarose. Additionally, this kit cannot be used to purify antibodies from serum because other serum proteins will also bind to the nickel column.

Removing non-conjugated enzyme using this kit can improve assay results. Increase in assay sensitivity, shorter washes and incubation times, and a high signal-to-noise ratio can be more easily achieved when free enzyme is absent from the system. Furthermore, the column concentrates samples, which eases assay optimization when using conjugates in dilute solutions.
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Labeling Scale0.5 to 3 mg
Product LinePierce
Product TypeConjugate Purification Kit
Quantity1 Kit
Sufficient For10 purifications, each of 0.5 to 50 mg of enzyme-conjugated antibody
Labeling TargetIgG
Label or DyeHRP (Horseradish Peroxidase)
Unit SizeEach
Contents & Storage
Sufficient For: 10 purifications, each of 0.5 to 50 mg of enzyme-conjugated antibody
  • IDA Agarose Column, 2 mL, 1 column
  • Polyacrylamide Desalting Column, 10 mL, 1 column
  • Column Activator Buffer, 25 mL
  • Elution Buffer, 35 mL
  • TBS Packs (each makes 500 mL), 2 packs
  • PBS Pack (makes 500 mL), 1 pack
  • Activity Preservation Solution, 1 mL

    Store kit at 4°C.