3'-Azido-3'-deoxythymidine, 98%
3'-Azido-3'-deoxythymidine, 98%
3'-Azido-3'-deoxythymidine, 98%
Thermo Scientific Chemicals

3'-Azido-3'-deoxythymidine, 98%

CAS: 30516-87-1 | C10H13N5O4 | 267.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 454980010
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.20
Save 6.55
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.20
Save 6.55
Each
Chemical Identifiers
CAS30516-87-1
IUPAC Name1-[(2R,4S,5S)-4-azido-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC10H13N5O4
InChI KeyHBOMLICNUCNMMY-CFQLRCIDNA-N
SMILESCC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point122°C to 126°C
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=97.5 %
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only