Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • OxPhos Rodent WB Antibodies

          Invitrogen

          OxPhos Rodent WB Antibody Cocktail

          22 Published Figures
          83 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite OxPhos Rodent WB Antibody Cocktail

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (22)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          OxPhos Rodent WB Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 23

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          OxPhos Rodent WB Antibody (45-8099) in WB

          Western blot analysis of OxPhos in Rat liver mitochondria using an OxPhos Monoclonal antibody cocktail (Product # 45-8099). The sample in lane 1 was kept at room temperature, whereas the remaining three samples were heated to 37°C, 50°C, and 100°C, respectively. This blot shows that boiling of samples leads to a decrease in signal due to aggregation of proteins therefore heating samples at or close to boiling is not recommended. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)
          OxPhos Rodent WB Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          45-8099

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 83 publications 83 publications
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Non-human primate, Rat

          Published species

          Human, Mouse, Pig, Rat, Sheep

          Host/Isotype

          Mouse / Ig

          Class

          Cocktail

          Type

          Antibody

          Clone

          Cocktail

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.5 mg/mL

          Amount

          300 µg

          Purification

          purified

          Storage buffer

          HEPES buffered saline

          Contains

          0.02% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533835

          Product Specific Information

          This product is a cocktail of multiple OxPhos antibodies.

          C-I-20 (ND6) (clone) 20E9DH10C12 - Bovine Complex I
          Purification Method: Antigen Affinity Chromatography

          C-II-30 (FeS) (clone) 21A11AE7 - Bovine Heart Complex II
          Purification Method: IgG Fraction

          C-III-Core 2 (clone) 13G12AF12BB11 - Bovine Heart Complex II, III
          Purification Method: IgG Fraction

          C-IV-I (clone) 1D6E1A8 - Human Complex IV subunit I
          Purification Method: Ammonium Sulfate Precipitation

          C-V-alpha (clone) 15H4C4 - Bovine Complex V
          Purification Method: Antigen Affinity Chromatography

          The kit is suitable for Western Blotting analysis of the relative levels of the 5 OxPhos complexes in mitochondrial preparations from mouse, rat, human, or bovine sources. These clones were chosen because they are against a subunit that is labile when its complex is not assembled. Moreover, the combination is readily resolved in SDS-PAGE when the appropriate gel conditions are used. Also, COXI is a highly hydrophobic protein and appears as a broad band at ~35 kDa (not at its true molecular weight at 57 kDa). It is very sensitive to heating. Therefore, the samples, including the positive control, should not be heated over 50°C before loaded on the gel.

          The antibody cocktail (1.5 mg/mL) should be diluted 250x to a final working concentration of 6.0 µg/mL for Western blotting. We recommend using PBS with 1% milk as the antibody diluent and using a high pH CAPS/PVDF transfer protocol.

          Target Information

          Altered levels of assembly can arise from mutations in individual subunits, mutations in assembly factors for the complex(es), mtDNA depletion or as a result of physiological and or pathological changes e.g. hormone treatment, exercise, diet or oxidative stress.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 13G12AF12BB11; 15H4C4; 1D6E1A8; 20E9DH10C12; 21A11AE7; OxPhos Complex

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-xmwlv:80/100.66.76.16:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline