Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NDUFA9 Antibodies

          Invitrogen

          NDUFA9 Monoclonal Antibody (20C11B11B11)

          Advanced Verification
          30 Published Figures
          80 References
          View all (13) NDUFA9 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NDUFA9 Monoclonal Antibody (20C11B11B11)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (30)
          • Advanced Verification (1)
          NDUFA9 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NDUFA9 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 33

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NDUFA9 Antibody (459100) in WB

          Western blot was performed using Anti-NDUFA9 Monoclonal Antibody (20C11B11B11) (Product # 45-9100) and a 36 kDa band corresponding to NDUFA9 was observed across cell lines and tissues tested. Membrane enriched cell extracts (30 µg lysate) of HeLa (Lane 1), Caco-2 (Lane 2), SW480 (Lane 3), Hep G2 (Lane 4), Jurkat (Lane 5), K-562 (Lane 6), MCF-7 (Lane 7), Mouse Liver (Lane 8), Rat Liver (Lane 9), Mouse Brain (Lane 10) and Rat Brain (Lane 11) were electrophoresed using Novex® NuPAGE® 12 % Bis-Tris gel (Product # NP0342BOX). Resolved proteins ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Immunocytochemistry (ICC/IF)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody in Western Blot (WB)
          NDUFA9 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          459100

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1.0 µg/mL
          View 74 publications 74 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          1-2 µg/10 ^6 cells
          -

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Rat

          Published species

          Horse, Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          20C11B11B11

          Immunogen

          Bovine Heart Complex I

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          IgG fraction

          Storage buffer

          HEPES buffered saline

          Contains

          0.02% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10376187

          Product Specific Information

          Purification is near homogeneity as judged by SDS-PAGE. The antibody was produced in vitro using hybridomas grown in serum-free medium, and then purified by biochemical fractionation.

          459100 detects a band at approximately 36 kDa.

          Use human heart mitochondria as a positive control.

          Target Information

          NDUFA9 (NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial) is also named as NDUFS2L, CI-39kD and belongs to the complex I NDUFA9 subunit family. It is involved in stabilizing the junction between membrane and matrix arms of complex I, a late assembly step critical for complex I biogenesis and activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CI-39kD; complex I 39kDa subunit; complex I-39kD; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa; NADH dehydrogenase (ubiquinone) 42 kDa subunit; NADH dehydrogenase (ubiquinone) Fe-S protein 2-like (NADH-coenzyme Q reductase); NADH-ubiquinone oxidoreductase 39 kDa subunit; short chain dehydrogenase/reductase family 22E, member 1; sperm flagella protein 3; unnamed protein product

          View more View less

          Gene Aliases: 1010001N11Rik; CC6; CI-39k; CI39k; COQ11; MC1DN26; NDUFS2L; SDR22E1

          View more View less

          UniProt ID: (Human) Q16795, (Bovine) P34943, (Mouse) Q9DC69, (Rat) Q5BK63

          View more View less

          Entrez Gene ID: (Human) 4704, (Bovine) 404188, (Mouse) 66108, (Rat) 362440

          View more View less

          Function(s)
          NADH dehydrogenase activity protein binding NADH dehydrogenase (ubiquinone) activity macromolecular complex binding oxidoreductase
          Process(es)
          mitochondrial electron transport, NADH to ubiquinone ubiquinone biosynthetic process sodium ion transport circadian rhythm aerobic respiration mitochondrial ATP synthesis coupled proton transport hydrogen ion transmembrane transport mitochondrial respiratory chain complex I assembly oxidation-reduction process response to glucose
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-nnlvt:80/100.66.74.151:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0