Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PGK1 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for will be discontinued once stock is depleted. Please check out our other {{productObj.targetName}} {{productObj.pendingDiscontinuationTargetLabel}} Please check out our other {{productObj.pendingDiscontinuationTargetLabel}} or contact Technical Support with any questions.

          Invitrogen

          PGK1 Monoclonal Antibody (22C5D8)

          99 Published Figures
          328 References
          View all (24) PGK1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Datasheet
          Certificates
          Protocols
          Datasheet
          Certificates
          Protocols

          Cite PGK1 Monoclonal Antibody (22C5D8)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Published Figures (99)
          PGK1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PGK1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 99

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PGK1 Antibody (459250) in WB

          Nucleic acids research 2018 - Figure 7. Rio1 responds to nutritional deprivation and crosstalks with Gcn4 at the transcriptional level. ( A ) The Rio1 transcriptome is enriched in transcripts of genes that are part of various stress-response systems. The main pie shows the percentage of the 818 Rio1-modulated genes that belong to specific stress transcriptomes (represented by the peripheral pies, which are size-weighted for the number... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)
          PGK1 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PGK1 Monoclonal Antibody (22C5D8)

          Product Details

          459250

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-1 µg/mL
          View 311 publications 311 publications

          Immunoprecipitation (IP)

          -
          View 2 publications 2 publications

          Dot blot (DB)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 14 publications 14 publications
          Product Specifications

          Published species

          Fungi, Human, Insect, Mouse, Yeast

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          22C5D8

          Immunogen

          Full-length native S. cerevisiae pGK protein (purified)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          250 µg

          Storage conditions

          4°C

          Shipping conditions

          Wet ice

          RRID

          AB_2532235

          Product Specific Information

          This product reacts with Saccharomyces cerevisiae PGK1 - predicted molecular weight: 45 kDa.

          Purity >95% by SDS-PAGE.

          Target Information

          Also known as ATP:3-phosphoglycerate 1-phosphotransferase showed that the plasmin reductase isolated from conditioned medium of fibrosarcoma cells is the glycolytic enzyme phosphoglycerate kinase. They concluded that phosphoglycerate kinase not only functions in glycolysis but is secreted by tumor cells and participates in the angiogenic process as a disulfide reductase.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch