Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • VDAC Antibodies

          Invitrogen

          Porin Monoclonal Antibody (16G9E6BC4)

          Advanced Verification
          42 Published Figures
          46 References
          View all (49) VDAC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Porin Monoclonal Antibody (16G9E6BC4)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (42)
          • Advanced Verification (1)
          Porin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Porin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 44

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Porin Antibody (459500) in WB

          Western blot analysis of Porin in isolated mitochondria using an NDUFA13 Monoclonal antibody (Product # 459500). Predicted band size: 30 kDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Porin Antibody in Western Blot (WB)
          Porin Antibody in Immunocytochemistry (ICC/IF)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody in Western Blot (WB)
          Porin Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Porin Monoclonal Antibody (16G9E6BC4)

          Product Details

          459500

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1.0 µg/mL
          View 40 publications 40 publications

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Immunomicroscopy (IM)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Yeast

          Published species

          Fungi, Human, Yeast

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          16G9E6BC4

          Immunogen

          Full length native protein (purified) corresponding to Saccharomyces cerevisiae VDAC1/Porin
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          IgG fraction

          Storage buffer

          HEPES buffered saline

          Contains

          0.02% sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2532239

          Target Information

          Voltage-dependent anion channel (VDAC) proteins are abundant, pore-forming proteins belonging to the eukaryotic mitochondrial porins. It was discovered in the mitochondrial outer membrane. It also is expressed in the plasma membrane. At least three different VDAC genes have been identified in vertebrates. VDAC proteins are known to play an essential role in cellular metabolism and in the early stages of apoptosis. For example, VDAC constitutes a major pathway by which metabolites such as ADP/ATP, succinate and citrate are exchanged between the cytosol and mitochondria. And VDAC1 in the plasma membrane establishes a novel level of apoptosis regulation putatively via its redox activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: hVDAC1; MGC111064; Mitochondrial Porin; PORIN-31-HL; VD

          View more View less

          Gene Aliases: OMP2; YNL055C

          View more View less

          UniProt ID: (Baker's yeast) P04840

          View more View less

          Entrez Gene ID: (Baker's yeast) 855669

          View more View less

          Function(s)
          voltage-gated anion channel activity porin activity ubiquinone binding voltage-gated ion channel
          Process(es)
          ion transport apoptotic process mitochondrion organization inorganic anion transport positive regulation of protein import into nucleus cell redox homeostasis DNA transport transmembrane transport positive regulation of protein serine/threonine kinase activity anion transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch