Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MTCO1 Antibodies

          Invitrogen

          MTCO1 Monoclonal Antibody (1D6E1A8)

          Advanced Verification
          53 Published Figures
          152 References
          View all (12) MTCO1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite MTCO1 Monoclonal Antibody (1D6E1A8)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (53)
          • Advanced Verification (2)
          MTCO1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MTCO1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 57

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MTCO1 Antibody (459600) in ICC/IF

          Immunofluorescence analysis of MTCO1 was performed using 70% confluent log phase HeLa cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with MTCO1 Monoclonal Antibody (1D6E1A8) (Product # 459600) at 5 µg/mL dilution in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor Plus 488 (Produc... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MTCO1 Antibody in Immunocytochemistry (ICC/IF)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MTCO1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          MTCO1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          MTCO1 Antibody in Immunocytochemistry (ICC/IF)
          MTCO1 Antibody in Immunocytochemistry (ICC/IF)
          MTCO1 Antibody in Immunocytochemistry (ICC/IF)
          MTCO1 Antibody in Immunocytochemistry (ICC/IF)
          MTCO1 Antibody in Immunocytochemistry (ICC/IF)
          MTCO1 Antibody in Immunocytochemistry (ICC/IF)
          MTCO1 Antibody in Immunocytochemistry (ICC/IF)
          MTCO1 Antibody in Immunocytochemistry (ICC/IF)
          MTCO1 Antibody in Immunocytochemistry (ICC/IF)
          MTCO1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          MTCO1 Antibody in Immunohistochemistry (IHC)
          MTCO1 Antibody in Immunohistochemistry (IHC)
          MTCO1 Antibody in Immunohistochemistry (IHC)
          MTCO1 Antibody in Immunohistochemistry (IHC)
          MTCO1 Antibody in Immunohistochemistry (IHC)
          MTCO1 Antibody in Immunohistochemistry (IHC)
          MTCO1 Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody in Western Blot (WB)
          MTCO1 Antibody
          MTCO1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          459600

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 93 publications 93 publications

          Immunohistochemistry (IHC)

          -
          View 14 publications 14 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:5,000
          View 4 publications 4 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 4 publications 4 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:100
          View 24 publications 24 publications

          Flow Cytometry (Flow)

          1:500
          -

          Miscellaneous PubMed (Misc)

          -
          View 12 publications 12 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Arthropod, Guinea pig, Human, Mouse, Rat, Zebrafish

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1D6E1A8

          Immunogen

          Human Mitochondrial Complex IV subunit I

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.99 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS with 0.05% BSA, 40% glycerol

          Contains

          0.01% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10374492

          Product Specific Information

          For best results with this antibody in Western blot, do not boil samples before loading onto the gel. Boiling of the sample will cause a loss of signal. Hydrophobic intrinsic membrane proteins such as the core mtDNA-encoded proteins of the mitochondrial OXPHOS complexes tend to run faster in SDS-PAGE than predicted by their amino acid composition. This is likely due to incomplete unfolding of the protein and a more negative charge:mass ratio. In Western blot applications, this antibody detects a band at approximately 40 kDa (predicted MW: 57 kDa). In mouse liver lysate a specific band below 37 kDa was detected.

          Recommended positive controls:

          ICC/IF - Pig retinal pigment epithelial cells, rat cerebellum primary cells

          IHC (P) - Human skeletal muscle and colon tissue, rat pancreas tissue, pig smooth muscle tissue

          WB - Mouse, human, bovine and rat heart mitochondria lysate

          Flow - HEK-293 cells

          Recommended long term storage at -20°C.

          Target Information

          Cytochrome c oxidase subunit I (CO1 or MTCO1) is 1 of 3 mitochondrial DNA (mtDNA) encoded subunits (MTCO1, MTCO2, MTCO3) of respiratory Complex IV. Complex IV is located within the mitochondrial inner membrane and is the third and final enzyme of the electron transport chain of mitochondrial oxidative phosphorylation. It is composed of 13 polypeptides. Subunits I, II, and III (MTCO1, MTCO2, MTCO3) are encoded by mtDNA while subunits IV, Va, Vb, VIa, VIb, VIc, VIIa, VIIb, VIIc, and VIII are nuclear encoded. The cytochrome c oxidase family of enzymes have 4 redox centers, 2 hemes and 2 copper centers. In mitochondrial Complex IV, the 2 hemes are a and a3 and the 2 coppers are CuA and CuB. The 2 hemes and CuB are bound to subunit I. Acin-Perez et al. (2003) identified a cell line containing single and double missense mutations in the cytochrome c oxidase (COX) subunit I gene of mouse mitochondrial DNA. And they hypothesized that deleterious mutations can arise and become predominant; cultured cells can maintain several mtDNA haplotypes at stable frequencies; the respiratory chain has little spare COX capacity; and that the size of a cavity in the vicinity of val421 in MTCO1I of animal COX may affect the function of the enzyme.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Co1; unnamed protein product

          View more View less

          Gene Aliases: CoxI

          View more View less

          Entrez Gene ID: (Mouse) 17708

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch