Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NKp46 (CD335) Antibodies

          Invitrogen

          CD335 (NKp46) Monoclonal Antibody (29A1.4), PerCP-eFluor™ 710, eBioscience™

          13 Published Figures
          30 References
          View all (52) NKp46 (CD335) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD335 (NKp46) Monoclonal Antibody (29A1.4), PerCP-eFluor™ 710, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (13)
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD335 (NKp46) Antibody (46-3351-80) in Flow

          Staining of BALB/c splenocytes with Anti-Mouse CD49b (Integrin alpha 2) PE (Product # 12-5971-82) and 0.125 µg of Rat IgG2a K Isotype Control PerCP-eFluor® 710 (Product # 46-4321-82) (left) or 0.125 µg of Anti-Mouse CD335 (NKp46) PerCP-eFluor® 710 (right). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)
          CD335 (NKp46) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          46-3351-80

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.25 µg/test
          View 30 publications 30 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), PerCP-eFluor™ 710, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          29A1.4

          Conjugate

          PerCP-eFluor™ 710 PerCP-eFluor™ 710 PerCP-eFluor™ 710
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant Violet 711
          • eFluor 450
          • eFluor 660
          • FITC
          • Functional Grade
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Request custom conjugation

          Excitation/Emission Max

          482/708 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1834442

          Product Specific Information

          Description: The monoclonal antibody 29A1.4 recognizes mouse NKp46, also known as CD335. CD335, a member of the natural cytotoxicity receptor (NCR) family, is a glycoprotein with 2 Ig-like domains and a short cytoplasmic tail. Expression of CD335 is uniquely found on NK cells (including immature NK cells, defined as DX5- CD3-, and thereby allowing discrimination between NKT cells and NK cells (NKp46+, CD3-). Furthermore, unlike many of the NK markers which also stain NKT cells, staining with 29A1.4 is not strain specific. Staining has been shown on C57Bl/6, SJL, CBA/CA and BALB/C strains. NKp46 has been shown to play a role in NK cell-mediated lysis of several tumor cells and pathogen-infected cell lines.

          The 29A1.4 monoclonal antibody has been shown to activate NK cells in vitro. The 29A1.4 monoclonal antibody does not deplete NK cells in vivo.

          Applications Reported: This 29A1.4 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This 29A1.4 antibody has been tested by flow cytometric analysis of mouse spleen cells. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative.

          Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols.

          Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD335, also known as NKp46 or NCR1, is a member of the natural cytotoxicity receptor family, which includes NKp30 and NKp44. This type I transmembrane glycoprotein is expressed on NK cells and plays a crucial role in activating and expanding resting NK cells through interactions with dendritic cells. CD335 enhances the ability of NK cells to eliminate tumor and virus-infected cells, such as those infected by influenza. CD335 consists of two Ig-like domains and is related to leukocyte immunoglobulin-like receptors (LIR) and killer inhibitory receptors (KIR). It contributes to the increased efficiency of activated NK cells in mediating tumor cell lysis. Additionally, CD335 interacts with pancreatic beta cells during the development of type 1 diabetes, highlighting its involvement in both immune defense and autoimmune processes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: activating receptor 1; hNKp46; Ly-94; lymphocyte antigen 94; mAR-1; MAR1 (mouse activating receptor 1); mNKp46; natural killer cell p46-related protein; natural killer cell receptor; NK-p46; unnamed protein product

          View more View less

          Gene Aliases: Cd335; Ly94; NKp46

          View more View less

          UniProt ID: (Mouse) Q8C567

          View more View less

          Entrez Gene ID: (Mouse) 17086

          View more View less

          Function(s)
          transmembrane signaling receptor activity protein binding immune receptor activity
          Process(es)
          immune response-inhibiting cell surface receptor signaling pathway detection of virus defense response to virus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch