eBioscience™ Streptavidin PerCP-eFluor™ 710 Conjugate, 25 μg - FAQs
eBioscience™ Streptavidin PerCP-eFluor™ 710 Conjugate, 25 μg - FAQs
View additional product information for eBioscience™ Streptavidin PerCP-eFluor™ 710 Conjugate - FAQs (46-4317-82, 46-4317-80)