Nur77 Monoclonal Antibody (12.14), PerCP-eFluor™ 710, eBioscience™ - FAQs
Nur77 Monoclonal Antibody (12.14), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Nur77 Monoclonal Antibody (12.14), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5965-82, 46-5965-80)