CCL3 (MIP-1 alpha) Monoclonal Antibody (DNT3CC), PerCP-eFluor™ 710, eBioscience™ - FAQs
CCL3 (MIP-1 alpha) Monoclonal Antibody (DNT3CC), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CCL3 (MIP-1 alpha) Monoclonal Antibody (DNT3CC), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7532-80, 46-7532-82)