c-MAF Monoclonal Antibody (sym0F1), PerCP-eFluor™ 710, eBioscience™ - FAQs
c-MAF Monoclonal Antibody (sym0F1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for c-MAF Monoclonal Antibody (sym0F1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9855-42, 46-9855-41)