Product Image
Thermo Scientific Chemicals

N-Benzoyladenosine, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
4646900101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 464690010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS4546-55-8
IUPAC NameN-{9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-6-yl}benzamide
Molecular FormulaC17H17N5O5
InChI KeyNZDWTKFDAUOODA-UCQOUBKUNA-N
SMILESOC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<5 %
HPLC>=95.0 %
Specific optical rotation-46° to -52° (20°C, 589 nm) (1% in DMSO)
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only