Sclareol
Thermo Scientific Chemicals

Sclareol

CAS: 515-03-7 | C20H36O2, C20H36O2 | 308.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 465120010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS515-03-7
IUPAC Name(1R,2R,8aS)-1-[(3R)-3-hydroxy-3-methylpent-4-en-1-yl]-2,5,5,8a-tetramethyl-decahydronaphthalen-2-ol
Molecular FormulaC20H36O2
InChI KeyXVULBTBTFGYVRC-FFADBYAMSA-N
SMILESC[C@@](O)(CC[C@H]1[C@](C)(O)CCC2C(C)(C)CCC[C@]12C)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and or chunks
Infrared spectrumConforms
Melting point99°C to 103°C
Appearance (Color)White to light yellow
GC>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only