Product Image
Thermo Scientific Chemicals

Afatinib

Have Questions?
Catalog NumberQuantity
465642500250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465642500
Price (USD)
108.65
Special offer
Online exclusive
Ends: 30-Jun-2026
128.00
Save 19.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
108.65
Special offer
Online exclusive
Ends: 30-Jun-2026
128.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS850140-72-6
IUPAC NameN-{4-[(3-chloro-4-fluorophenyl)amino]-7-(oxolan-3-yloxy)quinazolin-6-yl}-4-(dimethylamino)but-2-enamide
Molecular FormulaC24H25ClFN5O3
InChI KeyULXXDDBFHOBEHA-UHFFFAOYNA-N
SMILESCN(C)CC=CC(=O)NC1=C(OC2CCOC2)C=C2N=CN=C(NC3=CC(Cl)=C(F)C=C3)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=95.0 %
Infrared spectrumConforms
Appearance (Color)White to beige
Water=<5.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only