Moclobemide
Thermo Scientific Chemicals

Moclobemide

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465802500
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS71320-77-9
IUPAC Name4-chloro-N-[2-(morpholin-4-yl)ethyl]benzamide
Molecular FormulaC13H17ClN2O2
InChI KeyYHXISWVBGDMDLQ-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)C(=O)NCCN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.5 %
Loss on drying=<0.5 %
Appearance (Form)Crystalline powder
Appearance (Color)White to almost white
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only