DyLight™ 488 Maleimide
Thermo Scientific™

DyLight™ 488 Maleimide

Thermo Scientific DyLight 488 Sulfhydryl-Reactive Dye is a maleimide-activated derivative of high-performance DyLight 488 used to fluorescently label sulfhydryl-containing peptides,Read more
Have Questions?
Catalog NumberQuantity
466021 mg
Catalog number 46602
Price (USD)
465.00
Each
Quantity:
1 mg
Price (USD)
465.00
Each
Thermo Scientific DyLight 488 Sulfhydryl-Reactive Dye is a maleimide-activated derivative of high-performance DyLight 488 used to fluorescently label sulfhydryl-containing peptides, proteins, and other biomolecular probes.

DyLight 488 has high fluorescence intensity over a broad pH range (pH 4-9) and is more photostable than Cy2™ dye, Alexa Fluor™ 488, FITC, and LI-COR™ dyes in many applications. The high water solubility of DyLight Fluors allows a high dye-to-protein ratio to be achieved without causing precipitation of the conjugates.

Features of DyLight 488 Maleimide:

• High performance—DyLight 488 shows brighter fluorescence than Alexa Fluor 488, FITC and Cy2 dye
• Specific—maleimide-activated dye labels proteins and other molecules at reduced sulfhydryls (-SH)
• Efficient labeling methods—well-characterized chemistry and optimized protocols provide for reliable, high-quality labeling
• Optimized antibody labeling procedure—complete protocol for IgG reduction and labeling and calculating the labeling efficiency

Applications:
• Antibody labeling for immunofluorescence applications, including immunocytochemistry (ICC), immunohistochemistry (IHC), Western blotting and ELISA assay
• Target macromolecule labeling for in vitro and in vivo fluorescent detection strategies

DyLight 488 Sulfhydryl-Reactive Dye is activated with a maleic acid imide (maleimide) moiety to form a reactive alkylation reagent. Labeling occurs through reaction of the maleimide-activated dye with reduced sulfhydryl groups (-SH) to form stable thioether bonds. Maleimides are specific for sulfhydryl groups between pH 6.5–7.5. Learn more about maleimide chemistry.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityThiol
DescriptionDyLight 488 Maleimide
Emission518
Excitation493
Label or DyeDyLight™ 488
Product TypeDye
Quantity1 mg
Reactive MoietyMaleimide
SolubilityDMF (Dimethylformamide)
Label TypeDyLight Fluors
Product LineDyLight
Unit SizeEach
Contents & Storage
Store desiccated at -20°C. Shipped at ambient temperature.