DyLight™ 680 Maleimide
Thermo Scientific™

DyLight™ 680 Maleimide

Thermo Scientific DyLight 680 Sulfhydryl-Reactive Dye is a maleimide-activated derivative of high-performance DyLight 680 used to fluorescently label sulfhydryl-containing peptides,Read more
Have Questions?
Catalog NumberQuantity
466181 mg
Catalog number 46618
Price (USD)
344.65
Online exclusive
459.00
Save 114.35
Each
Quantity:
1 mg
Price (USD)
344.65
Online exclusive
459.00
Save 114.35
Each
Thermo Scientific DyLight 680 Sulfhydryl-Reactive Dye is a maleimide-activated derivative of high-performance DyLight 680 used to fluorescently label sulfhydryl-containing peptides, proteins and other biomolecular probes.

DyLight 680 produces near-infrared (IR) fluorescence that replaces other near-IR dyes, including Cy5.5™ dye and Alexa Fluor™ 680, and is ideal for multi-color applications. The high water solubility of DyLight Fluors means that a high dye-to-protein ratio can be attained without causing precipitation of the conjugates.

Features of DyLight 680 Maleimide:

• High performance—DyLight 680 fluoresces brighter than Alexa Fluor 680 and Cy5.5 dyes
• Specific—maleimide-activated dye labels proteins and other molecules at reduced sulfhydryls (-SH)
• Efficient labeling methods—well-characterized chemistry and optimized protocols provide for reliable, high-quality labeling
• Optimized antibody labeling procedure—complete protocol for IgG reduction and labeling and calculating the labeling efficiency

Applications:
• Antibody labeling for immunofluorescence applications, including immunocytochemistry (ICC), immunohistochemistry (IHC), Western blotting and ELISA assay
• Target macromolecule labeling for in vitro and in vivo fluorescent detection strategies

DyLight 680 Sulfhydryl-Reactive Dye is activated with a maleic acid imide (maleimide) moiety to form a reactive alkylation reagent. Labeling occurs through reaction of the maleimide-activated dye with reduced sulfhydryl groups (-SH) to form stable thioether bonds. Maleimides are specific for sulfhydryl groups between pH 6.5-7.5. Learn more about maleimide chemistry.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityThiol
DescriptionDyLight 680 Maleimide
Emission712
Excitation692
Label or DyeDyLight™ 680
Product TypeMaleimide
Quantity1 mg
Reactive MoietyMaleimide
SolubilityDMF (Dimethylformamide)
Label TypeDyLight Fluors
Product LineDyLight
Unit SizeEach
Contents & Storage
Store desiccated at -20°C.