Tazarotene
Thermo Scientific Chemicals

Tazarotene

Have Questions?
Catalog NumberQuantity
466302500250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466302500
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS118292-40-3
IUPAC Nameethyl 6-[2-(4,4-dimethyl-3,4-dihydro-2H-1-benzothiopyran-6-yl)ethynyl]pyridine-3-carboxylate
Molecular FormulaC21H21NO2S
InChI KeyOGQICQVSFDPSEI-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(N=C1)C#CC1=CC=C2SCCC(C)(C)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<1 %
HPLC>=97.5 %
Melting point102°C to 106°C
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only