Melanin, synthetic
Thermo Scientific Chemicals

Melanin, synthetic

CAS: 8049-97-6 | C18H10N2O4, C18H10N2O4 | 318.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466430010
Price (USD)
150.65
Special offer
Online exclusive
Ends: 28-Dec-2026
177.00
Save 26.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
150.65
Special offer
Online exclusive
Ends: 28-Dec-2026
177.00
Save 26.35
Each
Chemical Identifiers
CAS8049-97-6
IUPAC Name6,14-dimethyl-4,12-diazapentacyclo[8.6.1.1²,⁵.0¹³,¹⁷.0⁹,¹⁸]octadeca-1(17),2,5,9(18),10,13-hexaene-7,8,15,16-tetrone
Molecular FormulaC18H10N2O4
InChI KeyXUMBMVFBXHLACL-UHFFFAOYSA-N
SMILESCC1=C2NC=C3C2=C(C2=CNC4=C(C)C(=O)C(=O)C3=C24)C(=O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Nitrogen (N)6 to 7.5 %
Appearance (Color)Brown to black
Appearance (Form)powder
Infrared spectrumConforms
Carbon (C)44 to 55 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only