Pitavastatin calcium
Thermo Scientific Chemicals

Pitavastatin calcium

CAS: 147526-32-7 | C50H46CaF2N2O8, C50H46CaF2N2O8 | 881.00 g/mol
Have Questions?
Catalog NumberQuantity
466492500250 mg
Catalog number 466492500
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS147526-32-7
IUPAC Namecalcium bis((3R,5S,6E)-7-[2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl]-3,5-dihydroxyhept-6-enoate)
Molecular FormulaC50H46CaF2N2O8
InChI KeyRHGYHLPFVJEAOC-FFNUKLMVSA-L
SMILES[Ca++].O[C@H](C[C@H](O)C=CC1=C(N=C2C=CC=CC2=C1C1=CC=C(F)C=C1)C1CC1)CC([O-])=O.O[C@H](C[C@H](O)C=CC1=C(N=C2C=CC=CC2=C1C1=CC=C(F)C=C1)C1CC1)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<9 %
Specific optical rotation(20°C, 589 nm) (C=1.0, CH3CN,H2O (1:1))
Appearance (Form)Powder
Appearance (Color)White to yellow to beige
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only