Tiotropium bromide
Thermo Scientific Chemicals

Tiotropium bromide

CAS: 139404-48-1 | C19H22BrNO4S2, C19H22BrNO4S2 | 472.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 466520010
Price (USD)
653.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
653.00
Each
Chemical Identifiers
CAS139404-48-1
IUPAC Name(1R,2S,4R,5S)-7-{[2-hydroxy-2,2-bis(thiophen-2-yl)acetyl]oxy}-9,9-dimethyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-9-ium bromide
Molecular FormulaC19H22BrNO4S2
InChI KeyDQHNAVOVODVIMG-CJTHRXGHSA-M
SMILES[Br-].C[N+]1(C)[C@H]2CC(C[C@@H]1[C@@H]1O[C@H]21)OC(=O)C(O)(C1=CC=CS1)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=98.0 %
Water=<4.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only