Betulin
Thermo Scientific Chemicals

Betulin

CAS: 473-98-3 | C30H50O2, C30H50O2 | 442.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 466550050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS473-98-3
IUPAC Name(1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-3a-(hydroxymethyl)-5a,5b,8,8,11a-pentamethyl-1-(prop-1-en-2-yl)-icosahydro-1H-cyclopenta[a]chrysen-9-ol
Molecular FormulaC30H50O2
InChI KeyFVWJYYTZTCVBKE-ROUWMTJPSA-N
SMILESCC(=C)[C@@H]1CC[C@]2(CO)CC[C@]3(C)[C@H](CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)powder
Appearance (Color)White to light yellow to light beige
Infrared spectrumConforms
HPLC>=98.0 %
Loss on drying=<5.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only