Milnacipran hydrochloride
Milnacipran hydrochloride
Milnacipran hydrochloride
Thermo Scientific Chemicals

Milnacipran hydrochloride

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 466572500
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS101152-94-7
IUPAC Namehydrogen (1R,2S)-2-(aminomethyl)-N,N-diethyl-1-phenylcyclopropane-1-carboxamide chloride
Molecular FormulaC15H23ClN2O
InChI KeyXNCDYJFPRPDERF-PBCQUBLHSA-N
SMILES[H+].[Cl-].CCN(CC)C(=O)[C@@]1(C[C@@H]1CN)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point171°C to 181°C
Appearance (Color)White to beige
HPLC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only