Temozolomide
Thermo Scientific Chemicals

Temozolomide

CAS: 85622-93-1 | C6H6N6O2, C6H6N6O2 | 194.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466760050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS85622-93-1
IUPAC Name3-methyl-4-oxo-3H,4H-imidazo[4,3-d][1,2,3,5]tetrazine-8-carboxamide
Molecular FormulaC6H6N6O2
InChI KeyBPEGJWRSRHCHSN-UHFFFAOYSA-N
SMILESCN1N=NC2=C(N=CN2C1=O)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<2.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What is the recommended solubility of Temozolomide?

The solubility of temozolomide is 10 mg/mL in DMSO, which is what we recommend for reconstitution.