Temozolomide - FAQs
Temozolomide - FAQs
View additional product information for Temozolomide - FAQs (466760010, 466760050)
1 product FAQs found
What is the recommended solubility of Temozolomide?
The solubility of temozolomide is 10 mg/mL in DMSO, which is what we recommend for reconstitution.