Entinostat
Thermo Scientific Chemicals

Entinostat

CAS: 209783-80-2 | C21H20N4O3, C21H20N4O3 | 376.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 466870010
Price (USD)
726.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
726.00
Each
Chemical Identifiers
CAS209783-80-2
IUPAC Name(pyridin-3-yl)methyl N-({4-[(2-aminophenyl)carbamoyl]phenyl}methyl)carbamate
Molecular FormulaC21H20N4O3
InChI KeyINVTYAOGFAGBOE-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1NC(=O)C1=CC=C(CNC(=O)OCC2=CC=CN=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only