Telaprevir
Thermo Scientific Chemicals

Telaprevir

CAS: 402957-28-2 | C36H53N7O6, C36H53N7O6 | 679.86 g/mol
Have Questions?
Catalog NumberQuantity
4668900101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466890010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS402957-28-2
IUPAC Name3-{[(1S,3aR,6aS)-2-[(2S)-2-[(2S)-2-cyclohexyl-2-[(pyrazin-2-yl)formamido]acetamido]-3,3-dimethylbutanoyl]-octahydrocyclopenta[c]pyrrol-1-yl]formamido}-N-cyclopropyl-2-oxohexanamide
Molecular FormulaC36H53N7O6
InChI KeyBBAWEDCPNXPBQM-IGSHNTALSA-N
SMILESCCCC(NC(=O)[C@@H]1[C@H]2CCC[C@H]2CN1C(=O)[C@@H](NC(=O)[C@@H](NC(=O)C1=CN=CC=N1)C1CCCCC1)C(C)(C)C)C(=O)C(=O)NC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Appearance (Form)powder
HPLC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only