Cimetidine
Thermo Scientific Chemicals

Cimetidine

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467520250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS51481-61-9
IUPAC NameN-cyano-N''-methyl-N'-(2-{[(5-methyl-1H-imidazol-4-yl)methyl]sulfanyl}ethyl)guanidine
Molecular FormulaC10H16N6S
InChI KeyAQIXAKUUQRKLND-UHFFFAOYSA-N
SMILESCN=C(NCCSCC1=C(C)NC=N1)NC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point141°C to 143°C
Infrared spectrumConforms
Loss on drying=<0.5 %
Appearance (Form)Crystalline powder
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only