Tedizolid
Thermo Scientific Chemicals

Tedizolid

CAS: 856866-72-3 | C17H15FN6O3, C17H15FN6O3 | 370.34 g/mol
Have Questions?
Catalog NumberQuantity
4675800101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467580010
Price (USD)
269.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
269.00
Each
Chemical Identifiers
CAS856866-72-3
IUPAC Name3-{3-fluoro-4-[6-(2-methyl-2H-1,2,3,4-tetrazol-5-yl)pyridin-3-yl]phenyl}-5-(hydroxymethyl)-1,3-oxazolidin-2-one
Molecular FormulaC17H15FN6O3
InChI KeyXFALPSLJIHVRKE-UHFFFAOYNA-N
SMILESCN1N=NC(=N1)C1=NC=C(C=C1)C1=C(F)C=C(C=C1)N1CC(CO)OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
HPLC>=97.5 %
Loss on drying=<6 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only