Ligustrazine hydrochloride
Thermo Scientific Chemicals

Ligustrazine hydrochloride

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467640010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS76494-51-4
IUPAC Nametetramethylpyrazine hydrochloride
Molecular FormulaC8H13ClN2
InChI KeyRQKFOGXUTRDQPB-UHFFFAOYSA-N
SMILESCl.CC1=NC(C)=C(C)N=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
GC>=97.5 %
Titration Argentometric97.5 to 102.5 %
Infrared spectrumConforms
Appearance (Color)White to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only