Olopatadine hydrochloride
Thermo Scientific Chemicals

Olopatadine hydrochloride

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 467710050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS140462-76-6
IUPAC Namehydrogen 2-[(2Z)-2-[3-(dimethylamino)propylidene]-9-oxatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-5-yl]acetic acid chloride
Molecular FormulaC21H24ClNO3
InChI KeyHVRLZEKDTUEKQH-NOILCQHBSA-N
SMILES[H+].[Cl-].CN(C)CC\C=C1\C2=CC=CC=C2COC2=CC=C(CC(O)=O)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<0.5 % (105°C, 3 hr)
Appearance (Color)White to light yellow to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only