VE-821, 95%
Thermo Scientific Chemicals

VE-821, 95%

 
Have Questions?
Catalog NumberQuantity
467891000100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467891000
Price (USD)
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS1232410-49-9
IUPAC Name3-amino-6-(4-methanesulfonylphenyl)-N-phenylpyrazine-2-carboxamide
Molecular FormulaC18H16N4O3S
InChI KeyDUIHHZKTCSNTGM-UHFFFAOYSA-N
SMILESCS(=O)(=O)C1=CC=C(C=C1)C1=CN=C(N)C(=N1)C(=O)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Powder or solid
HPLC>=94.0 %
MScomplies
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only