SCH772984, 95%
Thermo Scientific Chemicals

SCH772984, 95%

 
Have Questions?
Catalog NumberQuantity
468271000100 mg
Catalog number 468271000
Price (USD)
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS942183-80-4
IUPAC Name1-(2-oxo-2-{4-[4-(pyrimidin-2-yl)phenyl]piperazin-1-yl}ethyl)-N-[3-(pyridin-4-yl)-1H-indazol-5-yl]pyrrolidine-3-carboxamide
Molecular FormulaC33H33N9O2
InChI KeyHDAJDNHIBCDLQF-UHFFFAOYNA-N
SMILESO=C(CN1CCC(C1)C(=O)NC1=CC=C2NN=C(C2=C1)C1=CC=NC=C1)N1CCN(CC1)C1=CC=C(C=C1)C1=NC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
MScomplies
NMR (1H-NMR)Consistent with structure
Appearance (Color)Yellow
Appearance (Form)Powder or solid
HPLC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only