CD62L (L-Selectin) Monoclonal Antibody (MEL-14), eFluor™ 450, eBioscience™ - FAQs
CD62L (L-Selectin) Monoclonal Antibody (MEL-14), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD62L (L-Selectin) Monoclonal Antibody (MEL-14), eFluor™ 450, eBioscience™ - FAQs (48-0621-80, 48-0621-82)