IL-2 Monoclonal Antibody (MQ1-17H12), eFluor™ 450, eBioscience™ - FAQs
IL-2 Monoclonal Antibody (MQ1-17H12), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IL-2 Monoclonal Antibody (MQ1-17H12), eFluor™ 450, eBioscience™ - FAQs (48-7029-42)