Random Primers - FAQs
Random Primers - FAQs
View additional product information for Random Primers - FAQs (48190011)
3 product FAQs found
Can I purchase Random Primers separately?
Yes, we do offer Random Primers as a stand-alone item: Cat. No. 48190011.
Which components of the SuperScript III First Strand Synthesis System for RT-PCR are available for purchase separately?
The following components are available as stand-alone items:
- Superscript III Reverse Transcriptase (Cat. Nos. 18080093, 18080044, 18080085)
- Oligo (dT)20 Primer (Cat. No. 18418020)
- Random hexamers (Cat. No. 48190011)
- 10 mM dNTP Mix (Cat. Nos. 18427013, 18427088)
- RNAseOUT Recombinant Ribonuclease Inhibitor (Cat. No. 10777019)
- E. coli RNAse H (Cat. Nos. 18021014, 18021071)
What is the concentration of Random Primers (Cat. No. 48190011)?
The concentration of Random Primers (Cat. No. 48190011) is 1500 pmol/µL or 1.5 mM.
Find additional tips, troubleshooting help, and resources within our Reverse Transcription and RACE Support Center.