Racks for Matrix™ 2D Barcoded Storage Tubes, 12mL screwtop - FAQs
Racks for Matrix™ 2D Barcoded Storage Tubes, 12mL screwtop - FAQs
View additional product information for Racks for Matrix™ 2D Barcoded Storage Tubes - FAQs (4905, 4890, 4893, 4896, 4897, 4898, 4899, 4900)
1 product FAQs found
Can I store the Thermo Scientific 2D barcoded storage tubes in standard 10x10 cryoboxes?
Yes, the Thermo Scientific 2D barcoded storage tubes can be stored in standard cryoboxes. However, we do not carry 2D barcode readers capable of reading a full box of 2D barcodes in this storage format.