Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Nanog Antibodies

          Invitrogen

          Nanog Monoclonal Antibody (eBioMLC-51), eFluor™ 660, eBioscience™

          75 Published Figures
          54 References
          View all (50) Nanog antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Nanog Monoclonal Antibody (eBioMLC-51), eFluor™ 660, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (75)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 76

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Nanog Antibody (50-5761-80) in ICC/IF

          Immunocytochemistry of fixed and permeabilized F9 cells stained with 10 µg/mL Rat IgG2a K Isotype Control eFluor® 660 (left) or 10 µg/mL Anti-Mouse Nanog eFluor® 660 (right). Nuclei are stained with DAPI; colocalization of signal appears pink. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunohistochemistry (IHC)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Immunocytochemistry (ICC/IF)
          Nanog Antibody in Western Blot, Immunocytochemistry, Flow Cytometry (WB, ICC/IF, Flow)
          Nanog Antibody in Immunohistochemistry (IHC)
          Nanog Antibody in Immunohistochemistry (IHC)
          Nanog Antibody in Immunohistochemistry (IHC)
          Nanog Antibody in Immunohistochemistry (IHC)
          Nanog Antibody in Immunohistochemistry (IHC)
          Nanog Antibody in Immunohistochemistry (IHC)
          Nanog Antibody in Immunohistochemistry (IHC)
          Nanog Antibody in Immunohistochemistry (IHC)
          Nanog Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Nanog Antibody in Western Blot (WB)
          Nanog Antibody in Western Blot (WB)
          Nanog Antibody in Western Blot (WB)
          Nanog Antibody in Western Blot (WB)
          Nanog Antibody in Western Blot (WB)
          Nanog Antibody in Western Blot (WB)
          Nanog Antibody in Western Blot (WB)
          Nanog Antibody in Western Blot (WB)
          Nanog Antibody in Flow Cytometry (Flow)
          Nanog Antibody in Flow Cytometry (Flow)
          Nanog Antibody in Flow Cytometry (Flow)
          Nanog Antibody in Flow Cytometry (Flow)
          Nanog Antibody in Flow Cytometry (Flow)
          Nanog Antibody in ChIP Assay (ChIP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          50-5761-80

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-Dependent
          View 8 publications 8 publications

          Immunohistochemistry (IHC)

          Assay-Dependent
          View 7 publications 7 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          10 µg/mL
          View 29 publications 29 publications

          Flow Cytometry (Flow)

          0.06 µg/test
          View 8 publications 8 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse, Rat

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 660, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBioMLC-51

          Conjugate

          eFluor™ 660 eFluor™ 660 eFluor™ 660
          • Unconjugated
          • Alexa Fluor 488 
          • Request custom conjugation

          Excitation/Emission Max

          651/668 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2574244

          Product Specific Information

          Description: The eBioMLC51 monoclonal antibody recognizes mouse Nanog. Nanog is a multidomain homeobox transcription factor that has been shown to maintain pluripotency of embryonic stem cells, independent of LIF/Stat3. Expression of Nanog in the mouse is specific to early embryos, the ICM of the blastocyst, embryonic stem (ES) cells, and embryonic germ (EG) cells. Nanog expression often overlaps, but is not identical to, that of Oct4. Nanog is downregulated upon cellular differentiation and loss of pluripotency, making it a suitable marker in determining the undifferentiated state of stem cells.

          Nanog acts as a transcriptional activator and has two activation domains in the C-terminus, called CD2 and WR, and one activation domain in the N terminus. The CD2 domain is unique to Nanog, whereas the NK2 DNA binding domain is well conserved. Immunoblotting using eBioMLC51 reveals a band at ~ 45 kDa in F9 (an embryonal carcinoma cell line) lysate, but not in lysate from the NIH3T3 cell line or mouse spleen. Preliminary data using fluorochrome-conjugated eBioMLC51 suggests that it is essential to use the eBioscience Foxp3 Staining Buffer Set, Product # 00-5523, for intracellular staining of mouse Nanog.

          Applications Reported: This eBioMLC-51 antibody has been reported for use in intracellular staining followed by flow cytometric analysis, western blotting, microscopy, immunohistochemical staining, and immunocytochemistry.

          Applications Tested: This eBioMLC-51 antibody has been tested by immunocytochemistry on fixed and permeabilized cells and can be used at less than or equal to 10 µg/mL. The eBioMLC-51 antibody has also been tested by intracellular staining followed by flow cytometric analysis of the F9 cell line using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol. This can be used at less than or equal to 0.06 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.

          Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          NANOG (Nanog homeobox) is a divergent homeodomain protein that directs pluripotency and differentiation of undifferentiated embryonic stem cells. NANOG mRNA is present in pluripotent mouse and human cell lines, and absent from differentiated cells. Human NANOG protein shares 52% overall amino acid identity with the mouse protein, and 85% identity in the homeodomain. Human NANOG maps to gene locus 12p13.31, while the mouse NANOG maps to gene loci 6 F2. Murine embryonic NANOG expression is detected in the inner cell mass of the blastocyst. Research studies have shown that high levels of human NANOG expression in the undifferentiated N-Tera embryonal carcinoma cell line. Further, NANOG is a transcription regulator involved in inner cell mass and embryonic stem (ES) cells proliferation and self-renewal. The role of NANOG in embryonic development suggested that it might be useful in the creation of stem cells that might be useful in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing POU5F1 (also known as Oct-4), another germline-specific transcription factor, and the transcription factors Sox2, Klf4 and Lin28 along with c-Myc in mouse fibroblasts. Experiments have demonstrated that iPS cells could be generated using expression plasmids expressing NANOG, Sox2, KlfF4 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine. When overexpressed, NANOG promotes cells to enter into S phase and proliferation. Diseases associated with dysfunction in the NANOG protein include tetracarcinoma and germ cell/embryonal cancer.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: early embryo specific expression NK family; early embryo specific expression NK-type homeobox protein; ES cell-associated protein 4; ES specific homeobox protein; FLJ12581; HGNC:20857; homeobox transcription factor Nanog; homeodomain protein; transcript variant 1a; transcript variant 1b; unnamed protein product

          View more View less

          Gene Aliases: 2410002E02Rik; ecat4; ENK; Stm1

          View more View less

          UniProt ID: (Mouse) Q80Z64

          View more View less

          Entrez Gene ID: (Mouse) 71950

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding protein binding transcription factor binding identical protein binding sequence-specific DNA binding sequence-specific double-stranded DNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter mesodermal cell fate commitment endodermal cell fate specification regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter positive regulation of cell proliferation gonad development embryonic pattern specification negative regulation of cell fate commitment gene expression stem cell division stem cell population maintenance cell differentiation negative regulation of BMP signaling pathway response to retinoic acid negative regulation of endodermal cell fate specification cell dedifferentiation negative regulation of cell differentiation positive regulation of transcription, DNA-templated positive regulation of mitotic cell cycle positive regulation of transcription from RNA polymerase II promoter stem cell differentiation regulation of stem cell division
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch