Nanog Monoclonal Antibody (eBioMLC-51), eFluor™ 660, eBioscience™ - FAQs
Nanog Monoclonal Antibody (eBioMLC-51), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Nanog Monoclonal Antibody (eBioMLC-51), eFluor™ 660, eBioscience™ - FAQs (50-5761-80, 50-5761-82)