Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Mouse IgM Antibodies

          Invitrogen

          Mouse IgM Monoclonal Antibody (II/41), eFluor™ 660, eBioscience™

          38 Published Figures
          33 References
          View all (37) Mouse IgM antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Mouse IgM Monoclonal Antibody (II/41), eFluor™ 660, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (38)
          Mouse IgM Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Mouse IgM Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 40

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Mouse IgM Antibody (50-5790-80) in ICC/IF

          Immunocytochemistry of fixed, differentiated OLN93 cells using 10 µg/mL of Anti-Oligodendrocyte Marker O1 Purified (Product # 14-6506-82) followed by 10 µg/mL of Anti-Mouse IgM eFluor® 660. Nuclei are stained with DAPI. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Mouse IgM Antibody in Immunocytochemistry (ICC/IF)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Immunocytochemistry (ICC/IF)
          Mouse IgM Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Mouse IgM Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          Mouse IgM Antibody in Immunohistochemistry (IHC)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry, ELISA (Flow, ELISA)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry, ELISA (Flow, ELISA)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in Flow Cytometry (Flow)
          Mouse IgM Antibody in ELISA (ELISA)
          Mouse IgM Antibody in ELISA (ELISA)
          Mouse IgM Antibody in ELISA (ELISA)
          Mouse IgM Antibody in ELISA (ELISA)
          Mouse IgM Antibody in T-Cell Activation (TCA)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          50-5790-80

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          10 µg/mL
          View 3 publications 3 publications

          Immunocytochemistry (ICC/IF)

          10 µg/mL
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          0.25 µg/test
          View 22 publications 22 publications

          ELISA (ELISA)

          -
          View 5 publications 5 publications

          T-Cell Activation (TCA)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Fish, Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 660, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          II/41

          Conjugate

          eFluor™ 660 eFluor™ 660 eFluor™ 660
          • Unconjugated
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant Violet 605
          • Brilliant Violet 711
          • eFluor 450
          • eFluor 570
          • FITC
          • NovaFluor Blue 725
          • NovaFluor Blue 800
          • NovaFluor Red 710
          • NovaFluor UV 765
          • NovaFluor Violet 690
          • NovaFluor Violet 745
          • NovaFluor Violet 800
          • NovaFluor Yellow 610
          • NovaFluor Yellow 810
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          651/668 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2574245

          Product Specific Information

          Description: The II/41 monoclonal antibody reacts with the mu heavy chain of mouse IgM. It does not react with other classes of mouse immunoglobulin including IgD, IgG or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells. Fluorochrome conjugated II/41 can be used as a detection secondary for mouse IgM.

          Applications Reported: This II/41 antibody has been reported for use in flow cytometric analysis, microscopy, immunohistochemical staining, and immunocytochemistry.

          Applications Tested: This II/41 antibody has been tested by immunocytochemistry on fixed cells and by immunohistochemistry on formalin-fixed paraffin embedded sections that were stained first with a primary antibody of the mouse IgM isotype. For immunocytochemistry and immunohistochemistry, this can be used at less than or equal to 10 µg/mL. This II/41 antibody has also been tested by flow cytometric analysis of mouse bone marrow cells and can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody by carefully titrated for optimal performance in the assay of interest.

          eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.

          Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Mouse IgM is an immunoglobulin M antibody that is produced by B cells in response to an antigen. It is the first antibody produced during an immune response and is the largest immunoglobulin in the serum. Mouse IgM is composed of five heavy chains and five light chains, and it is involved in complement activation and agglutination of antigens. It plays a crucial role in the innate immune response and is important for neutralizing pathogens before the adaptive immune response is fully activated. Mouse IgM is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: Ig heavy chain (V-D-J2) region; unnamed protein product; VH186.2-D-J-C mu

          View more View less

          Gene Aliases: Igh-6; Igh-M; Igh6; Igm; muH; TC1460681

          View more View less

          Entrez Gene ID: (Mouse) 16019

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch