Mouse IgM Monoclonal Antibody (II/41), eFluor™ 660, eBioscience™ - FAQs
Mouse IgM Monoclonal Antibody (II/41), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Mouse IgM Monoclonal Antibody (II/41), eFluor™ 660, eBioscience™ - FAQs (50-5790-82, 50-5790-80)