Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Ly-6G/Ly-6C Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for will be discontinued once stock is depleted. Please check out our other {{productObj.targetName}} {{productObj.pendingDiscontinuationTargetLabel}} Please check out our other {{productObj.pendingDiscontinuationTargetLabel}} or contact Technical Support with any questions.

          Invitrogen

          Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), eFluor™ 660, eBioscience™

          Advanced Verification
          96 Published Figures
          108 References
          View all (58) Ly-6G/Ly-6C antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Tech Support
          Datasheet
          Certificates
          Protocols
          Tech Support
          Datasheet
          Certificates
          Protocols
          Tech Support

          Cite Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), eFluor™ 660, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (96)
          • Advanced Verification (1)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 98

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Ly-6G/Ly-6C Antibody (50-5931-80) in Flow

          Staining of C57BL/6 bone marrow cells with Anti-Mouse CD11b FITC (Product # 11-0112-41) and 0.03 µg of Rat IgG2b K Isotype Control eFluor® 660 (Product # 50-4031-82) (left) or 0.03 µg of Anti-Mouse Ly-6G (Gr-1) eFluor® 660 (right). Cells in the large scatter population were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Immunocytochemistry (ICC/IF)
          Ly-6G/Ly-6C Antibody in Immunocytochemistry (ICC/IF)
          Ly-6G/Ly-6C Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Ly-6G/Ly-6C Antibody in Immunocytochemistry (ICC/IF)
          Ly-6G/Ly-6C Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Ly-6G/Ly-6C Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          50-5931-80

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 22 publications 22 publications

          Immunocytochemistry (ICC/IF)

          -
          View 6 publications 6 publications

          Flow Cytometry (Flow)

          0.06 µg/test
          View 74 publications 74 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Functional Assay (Functional)

          -
          View 3 publications 3 publications

          In Situ Hybridization (ISH)

          -
          View 1 publication 1 publication
          Product Specifications

          Published species

          Dog, Mouse

          Host/Isotype

          Rat / IgG2b, kappa

          Recommended Isotype Control

          Rat IgG2b kappa Isotype Control (eB149/10H5), eFluor™ 660, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          RB6-8C5

          Conjugate

          eFluor™ 660 eFluor™ 660 eFluor™ 660
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 561
          • Alexa Fluor 660 
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 563
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 650
          • Brilliant Violet 711
          • Brilliant Violet 786
          • eFluor 450
          • eFluor 506
          • FITC
          • Functional Grade
          • PE 
          • PE-Cyanine5
          • PE-Cyanine5.5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          651/668 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Wet ice

          RRID

          AB_2815189

          Product Specific Information

          Description: The RB6-8C5 monoclonal antibody reacts with mouse Ly-6G, a 21-25 kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations.

          eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (Product # 9668) be used when looking at Ly-6G specific targets.

          Applications Reported: This RB6-8C5 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This RB6-8C5 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.06 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.

          Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-lpswf:80/100.66.72.150:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0