Pdx1 Monoclonal Antibody (2A12), eFluor™ 660, eBioscience™ - FAQs
Pdx1 Monoclonal Antibody (2A12), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Pdx1 Monoclonal Antibody (2A12), eFluor™ 660, eBioscience™ - FAQs (50-6500-82)