IL-17A Monoclonal Antibody (eBio17B7), eFluor™ 660, eBioscience™ - FAQs
IL-17A Monoclonal Antibody (eBio17B7), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-17A Monoclonal Antibody (eBio17B7), eFluor™ 660, eBioscience™ - FAQs (50-7177-82, 50-7177-80)