Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IL-17A Antibodies

          Invitrogen

          IL-17A Monoclonal Antibody (eBio17B7), eFluor™ 660, eBioscience™

          96 Published Figures
          95 References
          View all (200) IL-17A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite IL-17A Monoclonal Antibody (eBio17B7), eFluor™ 660, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (96)
          IL-17A Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          IL-17A Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 97

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL-17A Antibody (50-7177-82) in Flow

          Mouse splenocytes were cultured for 7 days under Th17-polarization conditions. Cells were unstimulated (left) or stimulated (right) with Cell Stimulation Cocktail (plus protein transport inhibitors) (Product # 00-4975-03) and intracellularly stained with Anti-Mouse CD4 FITC (Product # 11-0041-82) and 0.125 µg of Anti-Mouse/Rat IL-17A eFluor® 660 using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Viable cells, as determined by Fixable Viability Dye eFluor® 780 (Product # 65-0865-14), were use... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Immunohistochemistry (IHC)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IL-17A Antibody in Immunohistochemistry (IHC)
          IL-17A Antibody in Immunohistochemistry (IHC)
          IL-17A Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          IL-17A Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          IL-17A Antibody in Immunohistochemistry (IHC)
          IL-17A Antibody in Immunohistochemistry (IHC)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry, ELISA (Flow, ELISA)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          50-7177-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-Dependent
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          0.25 µg/test
          View 85 publications 85 publications

          ELISA (ELISA)

          -
          View 3 publications 3 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 660, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBio17B7

          Conjugate

          eFluor™ 660 eFluor™ 660 eFluor™ 660
          • Alexa Fluor 488 
          • APC 
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 650
          • Brilliant Violet 711
          • Brilliant Violet 786
          • eFluor 450
          • eFluor 506
          • FITC
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • Request custom conjugation

          Excitation/Emission Max

          651/668 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11220280

          Product Specific Information

          Description: The eBio17B7 antibody reacts with mouse and rat IL-17A with no recognition of IL-17F. Interleukin-17A (IL-17A) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. The cDNA encoding human IL-17A was isolated from a library of CD4+ T cells; the encoded protein exhibits 72 percent amino acid identity with HVS13, an open reading frame from a T lymphotropic Herpesvirus saimiri, and 63 percent with mouse CTLA-8 (cytotoxic T-lymphocyte associated antigen-8). Human IL-17A exists as glycosylated 20-30 kD homodimers. High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells. IL-17A enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. Human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse IL-17A receptor.

          IL-23-dependent, IL-17A-producing CD4+ T cells (Th-17 cells) have been identified as a unique subset of Th cells that develops along a pathway that is distinct from the Th1- and Th2- cell differentiation pathways. The hallmark effector molecules of Th1 and Th2 cells, e.g., IFN gamma and IL-4, have each been found to negatively regulate the generation of these Th-17 cells.

          Applications Reported: This eBio17B7 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

          Applications Tested: This eBio17B7 antibody has been tested by intracellular staining and flow cytometric analysis of restimulated, Th17-polarized mouse splenocytes using the Intracellular Fixation and Permeabilization Buffer Set (Product # 88-8824-00) and protocol. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Staining has been successfully done using the Foxp3 buffer system (Product # 00-5523-00).

          eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.

          Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: cytotoxic T-lymphocyte-associated antigen 8; il 17; ILN; Interleukin; interleukin 17; Interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); Interleukin17A; R-IL-17-A; unnamed protein product

          View more View less

          Gene Aliases: CTLA-8; Ctla8; IL-17; IL-17A; Il17

          View more View less

          UniProt ID: (Mouse) Q62386

          View more View less

          Entrez Gene ID: (Mouse) 16171, (Rat) 301289

          View more View less

          Function(s)
          receptor binding cytokine activity protein binding protein homodimerization activity protein heterodimerization activity
          Process(es)
          positive regulation of antimicrobial peptide production inflammatory response Notch signaling pathway response to wounding gene expression positive regulation of keratinocyte proliferation keratinocyte differentiation positive regulation of interleukin-1 beta production positive regulation of interleukin-12 production positive regulation of interleukin-16 production positive regulation of interleukin-23 production positive regulation of interleukin-6 production positive regulation of tumor necrosis factor production interleukin-17A-mediated signaling pathway keratinocyte proliferation positive regulation of osteoclast differentiation positive regulation of transcription from RNA polymerase II promoter defense response to Gram-negative bacterium defense response to Gram-positive bacterium defense response to fungus positive regulation of protein kinase B signaling intestinal epithelial structure maintenance cellular response to interleukin-1 cellular response to glucocorticoid stimulus fibroblast activation interleukin-17-mediated signaling pathway granulocyte migration negative regulation of inflammatory response to wounding positive regulation of cytokine production involved in inflammatory response positive regulation of bicellular tight junction assembly negative regulation of interleukin-17-mediated signaling pathway positive regulation of chemokine (C-X-C motif) ligand 1 production regulation of gene expression response to amino acid
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch