Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • TNF alpha Antibodies

          Invitrogen

          TNF alpha Monoclonal Antibody (TN3-19.12), eFluor™ 660, eBioscience™

          7 Published Figures
          6 References
          View all (221) TNF alpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TNF alpha Monoclonal Antibody (TN3-19.12), eFluor™ 660, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (7)
          TNF alpha Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          TNF alpha Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TNF alpha Antibody (50-7423-82) in Flow

          Mouse splenocytes were unstimulated (left) or stimulated for 5 hours with Cell Stimulation Cocktail (plus protein transport inhibitors) (Product # 00-4975-03) (right), then intracellularly stained with Anti-Mouse CD4 PE (Product # 48-0041-82) and 0.25 µg of Anti-Mouse/Rat TNF alpha eFluor® 660. Total viable cells, as determined by Fixable Viability Dye eFluor® 780, were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TNF alpha Antibody in Flow Cytometry (Flow)
          TNF alpha Antibody in Western Blot (WB)
          TNF alpha Antibody in Flow Cytometry (Flow)
          TNF alpha Antibody in ELISA (ELISA)
          TNF alpha Antibody in ELISA (ELISA)
          TNF alpha Antibody in ELISA (ELISA)
          TNF alpha Antibody in Neutralization (Neu)
          TNF alpha Antibody in Neutralization (Neu)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          50-7423-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          0.5 µg/test
          View 2 publications 2 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Neutralization (Neu)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Rat

          Host/Isotype

          Armenian hamster / IgG

          Recommended Isotype Control

          Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 660, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          TN3-19.12

          Conjugate

          eFluor™ 660 eFluor™ 660 eFluor™ 660
          • Unconjugated
          • Biotin 
          • FITC
          • Functional Grade
          • PE 
          • PE-Cyanine7
          • Request custom conjugation

          Excitation/Emission Max

          651/668 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11219268

          Product Specific Information

          Description: The TN3-19.12 monoclonal antibody reacts with mouse, rat, and rabbit tumor necrosis factor alpha (TNF alpha), but not with human TNF alpha. The TN3-19.12 antibody is a neutralizing antibody. Mouse TNF alpha is a 17 kDa factor produced by macrophages, monocytes, neutrophils, CD4+ T cells and NK cells. A 26 kDa form of TNF alpha is expressed as a membrane-bound molecule. TNF alpha is cytolytic and plays an important role in immune regulation. Dimers and trimers of TNF alpha have been observed.

          Applications Reported: This TN3-19.12 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

          Applications Tested: This TN3-19.12 antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse splecnoytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.

          Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: cachectin; cytokine; DADB-70P7.1; pro-tumor necrosis factor; TNF alpha; TNF superfamily; TNF superfamily member 2; TNF α; TNFα; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor ligand 1f; tumor necrosis factor ligand superfamily member 2; tumor necrosis factor precursor; tumor necrosis factor-alpha; unnamed protein product

          View more View less

          Gene Aliases: DIF; RATTNF; TNF-a; TNF-alpha; Tnfa; TNFalpha; Tnfsf1a; TNFSF2; Tnlg1f

          View more View less

          UniProt ID: (Rat) P16599, (Mouse) P06804

          View more View less

          Entrez Gene ID: (Rat) 24835, (Mouse) 21926

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding protease binding cytokine activity tumor necrosis factor receptor binding identical protein binding receptor agonist activity histone H3K9ac reader activity protein binding death receptor agonist activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter response to hypoxia microglial cell activation positive regulation of cytokine production regulation of protein phosphorylation negative regulation of endothelial cell proliferation response to amphetamine negative regulation of L-glutamate transport macrophage activation involved in immune response chronic inflammatory response to antigenic stimulus regulation of immunoglobulin production positive regulation of immunoglobulin production positive regulation of immune effector process negative regulation of cytokine production involved in immune response positive regulation of chronic inflammatory response to antigenic stimulus positive regulation of humoral immune response mediated by circulating immunoglobulin skeletal muscle contraction negative regulation of systemic arterial blood pressure glucose metabolic process chromatin organization defense response inflammatory response immune response humoral immune response JNK cascade JAK-STAT cascade regulation of mitotic cell cycle circadian rhythm positive regulation of cell proliferation negative regulation of cell proliferation extrinsic apoptotic signaling pathway via death domain receptors intrinsic apoptotic signaling pathway in response to DNA damage response to xenobiotic stimulus response to mechanical stimulus response to virus response to bacterium response to fructose organ morphogenesis negative regulation of heart rate vascular endothelial growth factor production positive regulation of gene expression negative regulation of gene expression positive regulation of macrophage derived foam cell differentiation response to activity calcium-mediated signaling extracellular matrix organization osteoclast differentiation positive regulation of protein complex assembly positive regulation of defense response positive regulation of fever generation negative regulation of myelination lipopolysaccharide-mediated signaling pathway response to nutrient levels response to lipopolysaccharide negative regulation of interleukin-6 production positive regulation of chemokine production positive regulation of fractalkine production positive regulation of interferon-gamma production positive regulation of interleukin-1 beta production positive regulation of interleukin-18 production positive regulation of interleukin-6 production positive regulation of interleukin-8 production tumor necrosis factor-mediated signaling pathway positive regulation of heterotypic cell-cell adhesion toll-like receptor 3 signaling pathway response to isolation stress response to macrophage colony-stimulating factor regulation of cell proliferation vasodilation defense response to bacterium positive regulation of apoptotic process negative regulation of apoptotic process positive regulation of programmed cell death regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of I-kappaB kinase/NF-kappaB signaling response to amino acid positive regulation of protein complex disassembly positive regulation of MAPK cascade positive regulation of neuron apoptotic process negative regulation of blood vessel endothelial cell migration negative regulation of viral genome replication cellular extravasation positive regulation of nitric oxide biosynthetic process response to ethanol regulation of fat cell differentiation negative regulation of fat cell differentiation negative regulation of myoblast differentiation negative regulation of osteoblast differentiation regulation of osteoclast differentiation positive regulation of osteoclast differentiation positive regulation of protein catabolic process positive regulation of action potential positive regulation of cell adhesion positive regulation of mitotic nuclear division negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated negative regulation of mitotic cell cycle positive regulation of transcription from RNA polymerase II promoter positive regulation of translational initiation by iron negative regulation of glucose import positive regulation of JNK cascade response to antibiotic embryonic digestive tract development positive regulation of smooth muscle cell proliferation regulation of epithelial cell proliferation regulation of protein secretion regulation of inflammatory response regulation of developmental process regulation of insulin secretion modulation of synaptic transmission positive regulation of synaptic transmission regulation of synapse organization defense response to Gram-positive bacterium leukocyte migration leukocyte tethering or rolling detection of mechanical stimulus involved in sensory perception of pain negative regulation of lipid catabolic process positive regulation of nitric-oxide synthase activity positive regulation of membrane protein ectodomain proteolysis regulation of secretion positive regulation of protein transport response to glucocorticoid positive regulation of hair follicle development positive regulation of protein kinase B signaling positive regulation of glial cell proliferation positive regulation of vitamin D biosynthetic process epithelial cell proliferation involved in salivary gland morphogenesis regulation of branching involved in salivary gland morphogenesis negative regulation of vascular wound healing negative regulation of branching involved in lung morphogenesis positive regulation of calcineurin-NFAT signaling cascade cellular response to molecule of bacterial origin cellular response to lipopolysaccharide cellular response to amino acid stimulus cellular response to retinoic acid cellular response to nicotine cellular response to interferon-gamma cellular response to ionizing radiation positive regulation of podosome assembly endothelial cell apoptotic process protein localization to plasma membrane negative regulation of oxidative phosphorylation inflammatory response to wounding apoptotic signaling pathway extrinsic apoptotic signaling pathway cellular response to toxic substance liver regeneration necroptotic signaling pathway negative regulation of bile acid secretion antiviral innate immune response response to Gram-negative bacterium positive regulation of interleukin-33 production positive regulation of cytokine production involved in inflammatory response negative regulation of beta-amyloid clearance positive regulation of NIK/NF-kappaB signaling positive regulation of synoviocyte proliferation positive regulation of beta-amyloid formation response to L-glutamate positive regulation of neutrophil activation positive regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of protein localization to plasma membrane regulation of establishment of endothelial barrier negative regulation of bicellular tight junction assembly response to resveratrol cellular response to beta-amyloid positive regulation of vascular smooth muscle cell proliferation response to hydrogen sulfide positive regulation of leukocyte adhesion to vascular endothelial cell positive regulation of leukocyte adhesion to arterial endothelial cell response to quercetin response to 3,3',5-triiodo-L-thyronine response to gold nanoparticle positive regulation of protein localization to cell surface positive regulation of blood microparticle formation positive regulation of chemokine (C-X-C motif) ligand 2 production positive regulation of hepatocyte proliferation regulation of endothelial cell apoptotic process positive regulation of endothelial cell apoptotic process regulation of reactive oxygen species metabolic process positive regulation of DNA biosynthetic process positive regulation of estradiol secretion negative regulation of apoptotic signaling pathway positive regulation of extrinsic apoptotic signaling pathway negative regulation of extrinsic apoptotic signaling pathway in absence of ligand positive regulation of protein phosphorylation NIK/NF-kappaB signaling negative regulation of protein complex disassembly positive regulation of MAP kinase activity positive regulation of JUN kinase activity negative regulation of insulin receptor signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-wx6nt:80/100.66.72.150:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0