CXCL1 (GRO alpha) Monoclonal Antibody (W3ZLCXC), eFluor™ 660, eBioscience™ - FAQs
CXCL1 (GRO alpha) Monoclonal Antibody (W3ZLCXC), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CXCL1 (GRO alpha) Monoclonal Antibody (W3ZLCXC), eFluor™ 660, eBioscience™ - FAQs (50-7519-41, 50-7519-42)